Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Aspartic Acid (White Crystals or Crystalline Powder), Fisher BioReagents

Catalog No. BP374100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP374-100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP374-100

missing translation for 'validMainframeMsg'

This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-84-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00002616
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
Synonym l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid
PubChem CID 5960
ChEBI CHEBI:17053
IUPAC Name (2S)-2-aminobutanedioic acid
SMILES NC(CC(O)=O)C(O)=O

Specifications

Melting Point 270°C
Ammonium 0.02% max.
pH 2.5 to 3.5
Color White
Heavy Metals (as Pb) 10ppm max.
Ignition Residue 0.1% max.
Identification Pass Test
Loss on Drying 0.2% max.
Assay Percent Range 98.5 to 101.0 %
Other Amino Acids Chromatographically not detectable
Packaging Glass Bottle
Specific Rotation 24.8 to 25.8° (+ or -)
Merck Index 15, 830
State of Solution (%T) 98.0% min.
Alpha Vector Aspartic Acid
Density 1.6613g/cm³
Quantity 100 g
Formula Weight 133.05
Percent Purity 98.5 to 101.0%
Physical Form Solid
Chemical Name or Material L-Aspartic Acid
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.