Learn More
Medchemexpress LLC L-Aspartyl-L-phenylalanine | 13433-09-5 | 98.0% | 280.28 | 1 ML
SDP
Catalog No. 5000402565
Encompass_Preferred
L-Aspartyl-L-phenylalanine is a metabolite of aspartame that can inhibit angiotensin converting enzyme (ACE) purified from rabbit lungs with a Ki of 11 μM. It is suitable for laboratory research applications.
  • Inhibits angiotensin converting enzyme (ACE)
  • Metabolite of aspartame
  • Suitable for research use only
Catalog No. 50-004-02565 Supplier Medchemexpress LLC Supplier No. HY2309310MM/1ML
L-Aspartyl-L-phenylalanine is a metabolite of aspartame that can inhibit angiotensin converting enzyme (ACE) purified from rabbit lungs with a Ki of 11 μM. It is suitable for laboratory research applications.
  • Inhibits angiotensin converting enzyme (ACE)
  • Metabolite of aspartame
  • Suitable for research use only