Learn More
Medchemexpress LLC L-Aspartyl-L-phenylalanine | 13433-09-5 | 98.0% | 280.28 | 5 G
SDP
Catalog No. 5000400450
Encompass_Preferred
L-Aspartyl-L-phenylalanine is a metabolite of aspartame that effectively inhibits angiotensin converting enzyme (ACE) purified from rabbit lungs. It is provided as a laboratory chemical for research purposes only.
  • ACE inhibitor
  • For research use only
Catalog No. 50-004-00450 Supplier Medchemexpress LLC Supplier No. HY230935G
L-Aspartyl-L-phenylalanine is a metabolite of aspartame that effectively inhibits angiotensin converting enzyme (ACE) purified from rabbit lungs. It is provided as a laboratory chemical for research purposes only.
  • ACE inhibitor
  • For research use only