Learn More
Cayman Chemical L-Aspartyl-L-phenylalnIn 250mg
SDP
Supplier:  Cayman Chemical 34197250 MG
Encompass_Preferred
An ACE inhibitor and aspartame metabolite; an ACE inhibitor (Ki = 11 µM); serum levels are positively associated with pancreatic ductal adenocarcinoma
Catalog No. 50-294-8063