Learn More
Medchemexpress LLC HY-13735C 10mg Medchemexpress, l-Atabrine dihydrochloride CAS:56100-42-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13735C10MG
Encompass_Preferred
Medchemexpress, HY-13735C 10mg l-Atabrine dihydrochloride CAS:56100-42-6 l-Atabrine dihydrochloride is a less active enantiomer of quinacrine which displays antiprion activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2683