Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC l-Atabrine dihydrochloride | 56100-42-6 | MFCD31630799 | 98.4% | 472.88 | C23H32Cl3N3O | 5 MG
SDP

Catalog No. 5000222418
Encompass_Preferred

l-Atabrine dihydrochloride is the levorotatory enantiomer of quinacrine that exhibits antiprion activity. It is provided for research use in solid form and as a DMSO solution, and is intended for biochemical and prion biology studies requiring a well-characterized acridine derivative.

  • High purity (98.35%).
  • Molecular weight 472.88 and formula C23H32Cl3N3O.
  • Available as solid and as 10 mM solution in DMSO.
  • Suitable for prion research and biochemical assays.
  • Datasheet and safety data sheet available to support handling and storage.

Catalog No. 50-002-22418 Supplier Medchemexpress LLC Supplier No. HY13735C5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

l-Atabrine dihydrochloride is the levorotatory enantiomer of quinacrine that exhibits antiprion activity. It is provided for research use in solid form and as a DMSO solution, and is intended for biochemical and prion biology studies requiring a well-characterized acridine derivative.

  • High purity (98.35%).
  • Molecular weight 472.88 and formula C23H32Cl3N3O.
  • Available as solid and as 10 mM solution in DMSO.
  • Suitable for prion research and biochemical assays.
  • Datasheet and safety data sheet available to support handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.