Learn More
Sigma Aldrich Fine Chemicals Biosciences L Azetidine 2 carboxylic a1G
Catalog No. 502907516
Encompass
L-Azetidine-2-carboxylic acid is a non-protein amino acid and teratogenic agent. It is toxic in nature.
Catalog No. 50-290-7516 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A07601G
L-Azetidine-2-carboxylic acid is a non-protein amino acid and teratogenic agent. It is toxic in nature.