Learn More
Medchemexpress LLC L-Azetidine-2-carboxylic acid | 2133-34-8 | 99.9% | 101.10 | 500 MG
SDP
Catalog No. 5000365884
Encompass_Preferred
L-Azetidine-2-carboxylic acid is a proline analog that influences cellular processes such as lipid autophagy and endoplasmic reticulum stress. It is known to upregulate the lipid autophagy marker LC3-II and increase pro-apoptotic BAX protein, while also inducing ATF6 cleavage and upregulating phosphorylated eIF2α levels. This compound induces ER stress, leading to protein misfolding and aggregation, and has demonstrated teratogenic, pro-inflammatory, and pro-apoptotic effects.
  • Upregulates the lipid autophagy marker LC3-II via activation of the PERK pathway.
  • Increases pro-apoptotic BAX protein.
  • Induces ATF6 cleavage and upregulates phosphorylated eIF2α levels.
  • Induces ER stress, leading to protein misfolding and aggregation.
  • Shows teratogenic, pro-inflammatory, and pro-apoptotic effects.
Catalog No. 50-003-65884 Supplier Medchemexpress LLC Supplier No. 500MGHYW050044
L-Azetidine-2-carboxylic acid is a proline analog that influences cellular processes such as lipid autophagy and endoplasmic reticulum stress. It is known to upregulate the lipid autophagy marker LC3-II and increase pro-apoptotic BAX protein, while also inducing ATF6 cleavage and upregulating phosphorylated eIF2α levels. This compound induces ER stress, leading to protein misfolding and aggregation, and has demonstrated teratogenic, pro-inflammatory, and pro-apoptotic effects.
  • Upregulates the lipid autophagy marker LC3-II via activation of the PERK pathway.
  • Increases pro-apoptotic BAX protein.
  • Induces ATF6 cleavage and upregulates phosphorylated eIF2α levels.
  • Induces ER stress, leading to protein misfolding and aggregation.
  • Shows teratogenic, pro-inflammatory, and pro-apoptotic effects.