Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-azetidine-2-carboxylic acid | 2133-34-8 | MFCD00005166 | 99.9% | 101.10 g/mol | C4H7NO2 | 1 G
SDP

Catalog No. 5000209896
Encompass_Preferred

L-azetidine-2-carboxylic acid is a non-proteinogenic amino acid and a proline analog used in biochemical and cell biology research to induce endoplasmic reticulum (ER) stress, protein misfolding, and related cellular pathways. It is provided as a solid for laboratory use and includes documentation such as COA and SDS.

  • Non-proteinogenic amino acid proline analog.
  • CAS number 2133-34-8.
  • Molecular formula C4H7NO2.
  • Molecular weight 101.10 g/mol.
  • Purity 99.9%.
  • Appearance solid, white to off-white.
  • Soluble in water (100 mg/mL with ultrasonic assistance); sparingly soluble in DMSO.
  • Storage 4°C, protect from light; in solution -80°C (6 months) or -20°C (1 month).
  • Available pack sizes include 1 G.

Catalog No. 50-002-09896 Supplier Medchemexpress LLC Supplier No. HYW0500441G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-azetidine-2-carboxylic acid is a non-proteinogenic amino acid and a proline analog used in biochemical and cell biology research to induce endoplasmic reticulum (ER) stress, protein misfolding, and related cellular pathways. It is provided as a solid for laboratory use and includes documentation such as COA and SDS.

  • Non-proteinogenic amino acid proline analog.
  • CAS number 2133-34-8.
  • Molecular formula C4H7NO2.
  • Molecular weight 101.10 g/mol.
  • Purity 99.9%.
  • Appearance solid, white to off-white.
  • Soluble in water (100 mg/mL with ultrasonic assistance); sparingly soluble in DMSO.
  • Storage 4°C, protect from light; in solution -80°C (6 months) or -20°C (1 month).
  • Available pack sizes include 1 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.