Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Azetidine-2-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. A1043100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1043100MG 100 mg
A10431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1043100MG Supplier TCI America Supplier No. A1043100MG
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2133-34-8
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.105
MDL Number MFCD00005166
InChI Key IADUEWIQBXOCDZ-VKHMYHEASA-N
Synonym l-azetidine-2-carboxylic acid, s-azetidine-2-carboxylic acid, s---2-azetidinecarboxylic acid, 2s-azetidine-2-carboxylic acid, azetidyl-2-carboxylic acid, l-azetidine 2-carboxylic acid, s-2-azetidinecarboxylic acid, 2-azetidinecarboxylic acid, s, s---azetidine-2-carboxylic acid, 2-azetidinecarboxylic acid, l
PubChem CID 16486
ChEBI CHEBI:6198
IUPAC Name (2S)-azetidine-2-carboxylic acid
SMILES C1CNC1C(=O)O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 101.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Azetidine-2-carboxylic Acid [Antagonist of L-Proline]
RTECSNumber CM4310500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.