Learn More
Medchemexpress LLC HY-140346A 50mg Medchemexpress, L-Azidohomoalanine (hydrochloride) CAS:942518-29-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY140346A50MG
Encompass_Preferred
Medchemexpress, HY-140346A 50mg L-Azidohomoalanine (hydrochloride) CAS:942518-29-8 L-Azidohomoalanine hydrochloride is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1551