Learn More
TARGETMOL CHEMICALS INC L-Azidohomoalanine hydrochlori
SDP
Catalog No. 502878837
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. L-Azidohomoalanine hydrochloride (H-L-Aha-OH hydrochloride) is an alkyl chain-based PROTAC linker. Purity 98.16%
Catalog No. 50-287-8837 Supplier TARGETMOL CHEMICALS INC Supplier No. T41090100MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. L-Azidohomoalanine hydrochloride (H-L-Aha-OH hydrochloride) is an alkyl chain-based PROTAC linker. Purity 98.16%