Learn More
Medchemexpress LLC HY-131033 10mg Medchemexpress, L-Azidonorleucine (hydrochloride) CAS:1454334-76-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13103310MG
Encompass_Preferred
Medchemexpress, HY-131033 10mg L-Azidonorleucine (hydrochloride) CAS:1454334-76-9 L-Azidonorleucine hydrochloride, an unnatural amino acid, is A Methionine surrogate. L-Azidonorleucine hydrochloride can be used to label mammalian cell proteins and identify a diverse set of methionyl-tRNA synthetase (MetRS) mutants . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1257