Learn More
Medchemexpress LLC L-azidonorleucine (hydrochloride) | 1454334-76-9 | 98.0% | 200 MG
SDP
Catalog No. 5000340675
Encompass_Preferred
L-Azidonorleucine hydrochloride is an unnatural amino acid that serves as a methionine surrogate. It is used to label mammalian cell proteins and identify various methionyl-tRNA synthetase (MetRS) mutants. This compound functions as a click chemistry reagent due to its azide group, which enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Unnatural amino acid and methionine surrogate
  • Labels mammalian cell proteins
  • Identifies methionyl-tRNA synthetase (MetRS) mutants
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-40675 Supplier Medchemexpress LLC Supplier No. HY131033200MG
L-Azidonorleucine hydrochloride is an unnatural amino acid that serves as a methionine surrogate. It is used to label mammalian cell proteins and identify various methionyl-tRNA synthetase (MetRS) mutants. This compound functions as a click chemistry reagent due to its azide group, which enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Unnatural amino acid and methionine surrogate
  • Labels mammalian cell proteins
  • Identifies methionyl-tRNA synthetase (MetRS) mutants
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions