Learn More
Bachem Bz-Cit-OMe
SDP
Supplier:  Bachem 4014973.0005
Encompass_Preferred
Bz-Cit-OMe, 14325-36-1, C14H19N3O4, Mr 293.32, 5g. Bz-Orn(carbamoyl)-OMe. L-BCME, uncharged analog of the trypsin substrate L-BAME, is cleaved by human urinary arginine esterase-2 and trypsin.
Catalog No. 50-194-7136