Learn More
Enzo Life Sciences L-Biopterin (100mg). CAS: 22150-76-1
SDP
Catalog No. 502009267
Encompass_Preferred
Alternative name: 6-Biopterin, 2-Amino-6-[(1R,2S)-1,2-dihydroxypropyl]-4(1H)-pteridinone, 2-Amino-4-hydroxy-6-(1,2-dihydroxypropyl)pteridine, L-erythro-6-(1,2-Dihydroxypropyl)pterin. Purity: ≥98% (HPLC). Solubility: Soluble in 1 N sodium hydroxide or 1N hydrogen chloride; very slightly soluble in water (0.7mg/ml) (ultrasonication). To obtain concentrated solutions, ammonium hydroxide should be added. Long Term Storage: -20°C.
Catalog No. 50-200-9267 Supplier Enzo Life Sciences Supplier No. ALX440008M100
Add to Cart
Add to Cart
Alternative name: 6-Biopterin, 2-Amino-6-[(1R,2S)-1,2-dihydroxypropyl]-4(1H)-pteridinone, 2-Amino-4-hydroxy-6-(1,2-dihydroxypropyl)pteridine, L-erythro-6-(1,2-Dihydroxypropyl)pterin. Purity: ≥98% (HPLC). Solubility: Soluble in 1 N sodium hydroxide or 1N hydrogen chloride; very slightly soluble in water (0.7mg/ml) (ultrasonication). To obtain concentrated solutions, ammonium hydroxide should be added. Long Term Storage: -20°C.