Learn More
Medchemexpress LLC L-biotin | 21788-37-4 | 99.7% | 244.31 g/mol | C10H16N2O3S | 10 MG
SDP
Catalog No. 5000210072
Encompass_Preferred
L-biotin is the unnatural enantiomer of biotin provided as a research-grade chemical standard for stereochemical studies and analytical applications. It is supplied with a certificate of analysis and a safety data sheet to support safe handling and quality verification.
  • high reported purity suitable for analytical use
  • available in small milligram pack sizes for research
  • comes with COA and SDS documentation
  • chemical formula C10H16N2O3S and molecular weight 244.31 g/mol
  • intended for research use only; not for human use
Catalog No. 50-002-10072 Supplier Medchemexpress LLC Supplier No. HYW04844910MG
L-biotin is the unnatural enantiomer of biotin provided as a research-grade chemical standard for stereochemical studies and analytical applications. It is supplied with a certificate of analysis and a safety data sheet to support safe handling and quality verification.
  • high reported purity suitable for analytical use
  • available in small milligram pack sizes for research
  • comes with COA and SDS documentation
  • chemical formula C10H16N2O3S and molecular weight 244.31 g/mol
  • intended for research use only; not for human use