Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-biotin | 21788-37-4 | 99.7% | 244.31 g/mol | C10H16N2O3S | 50 MG
SDP

Catalog No. 5000210861
Encompass_Preferred

L-biotin is the unnatural enantiomer of biotin supplied as a high-purity research reagent for laboratory use (not for human or clinical applications).

  • High purity (reported 99.5-99.7%).
  • Cas number 21788-37-4.
  • Molecular weight 244.31 g/mol.
  • Chemical formula C10H16N2O3S.
  • Available in small milligram quantities such as 50 MG.

Catalog No. 50-002-10861 Supplier Medchemexpress LLC Supplier No. HYW04844950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-biotin is the unnatural enantiomer of biotin supplied as a high-purity research reagent for laboratory use (not for human or clinical applications).

  • High purity (reported 99.5-99.7%).
  • Cas number 21788-37-4.
  • Molecular weight 244.31 g/mol.
  • Chemical formula C10H16N2O3S.
  • Available in small milligram quantities such as 50 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.