Learn More
Medchemexpress LLC L-biotin (unnatural enantiomer of biotin) | 21788-37-4 | MFCD22373639 | 99.5% | 244.31 g·mol⁻¹ | C10H16N2O3S | 1 ML
SDP
Catalog No. 5000210074
Encompass_Preferred
L-biotin is the unnatural enantiomer of biotin supplied as a research reagent. It binds streptavidin in vitro and is provided as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid powder in multiple pack sizes. Recommended storage conditions are -20°C for powder, and -80°C for stock solutions (up to 6 months).
  • Unnatural enantiomer of biotin
  • Binds streptavidin in vitro
  • Available as 10 mM DMSO solution or solid powder
  • High purity (>99%)
  • Soluble in DMSO at high concentration
  • Powder storage: -20°C; stock solution storage: -80°C (6 months)
Catalog No. 50-002-10074 Supplier Medchemexpress LLC Supplier No. HYW04844910MM/1ML
L-biotin is the unnatural enantiomer of biotin supplied as a research reagent. It binds streptavidin in vitro and is provided as a ready-to-use 10 mM solution in DMSO (1 mL) or as solid powder in multiple pack sizes. Recommended storage conditions are -20°C for powder, and -80°C for stock solutions (up to 6 months).
  • Unnatural enantiomer of biotin
  • Binds streptavidin in vitro
  • Available as 10 mM DMSO solution or solid powder
  • High purity (>99%)
  • Soluble in DMSO at high concentration
  • Powder storage: -20°C; stock solution storage: -80°C (6 months)