Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-(-)-Bornyl acetate | 5655-61-8 | 196.29 | 1 ML
SDP

Catalog No. 5000416295
Encompass_Preferred

L-(-)-Bornyl acetate is supplied as a liquid, primarily used as a laboratory chemical and for the manufacture of substances. This compound is stable under recommended storage conditions.

  • Appearance as a colorless to light yellow liquid.
  • Stable under recommended storage conditions.
  • Container should be kept tightly sealed in a cool, well-ventilated area.
  • Keep away from direct sunlight and sources of ignition.
  • Recommended storage for pure form is -20°C for 3 years or 4°C for 2 years.
  • Recommended storage in solvent is -80°C for 6 months or -20°C for 1 month.
  • Shipping at room temperature if less than 2 weeks.

Catalog No. 50-004-16295 Supplier Medchemexpress LLC Supplier No. HYN0756A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-(-)-Bornyl acetate is supplied as a liquid, primarily used as a laboratory chemical and for the manufacture of substances. This compound is stable under recommended storage conditions.

  • Appearance as a colorless to light yellow liquid.
  • Stable under recommended storage conditions.
  • Container should be kept tightly sealed in a cool, well-ventilated area.
  • Keep away from direct sunlight and sources of ignition.
  • Recommended storage for pure form is -20°C for 3 years or 4°C for 2 years.
  • Recommended storage in solvent is -80°C for 6 months or -20°C for 1 month.
  • Shipping at room temperature if less than 2 weeks.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.