Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ L-Broth Agar (Capsules)
GSA_VA

Catalog No. MP113001241 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113001241 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP113001241 Supplier MP Biomedicals™ Supplier No. 113001241
Add to Cart
Add to Cart

Packaged in a capsule for convenience and ease of use. MP Biomedicals L-Broth Agar (Capsules) has the same high-quality, nutritionally rich formulation with the same growth characteristics as L-Broth agar powder, but in a capsule form that eliminates the

This nutritionally rich medium was originally developed for growth and maintenance of E. coli strains. The capsule format eliminates the waste, inaccuracies, and mess associated with weighing out bulk powder for the ultimate in ease and convenience. Tryptone and yeast extracts supply essential growth factors that E. coli cells would otherwise have to synthesize. The medium also contains essential electrolytes for transport and osmotic balance, due to the NaCl component.

  • Reagents are tested and chosen for their ability to promote optimum growth of E. coli
  • Chosen reagents are combined in the proper proportions according to the L-Broth Agar formulation and thoroughly blended for a uniform distribution
  • Samples of the blended formulation are used to prepare agar medium
  • The combinations are mixed to dissolve components and then autoclaved at 121°C for 15 minutes
  • Recommended use is 31 capsules/liter of water

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Description L-broth agar
Format Capsule
For Use With (Application) Bacterial culture
Quantity 5 kg
Regulatory Status RUO
Specimen Type E. coli
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.