Learn More
Teknova LB Luria Broth. 1000mL, Sterile
SDP
Catalog No. 50842651
Encompass
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth of bacteria. It is also known as Luria broth or Luria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli.
This item is not returnable. View return policy
Catalog No. 50-842-651 Supplier Teknova Supplier No. L8600
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth of bacteria. It is also known as Luria broth or Luria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli.