Learn More
Bellco Glass, Inc. L Bubbler Tube, 15L
SDP
Catalog No. 501944804
Encompass_Preferred
Sparging accessory for vertical sidearm spinner flasks. Manufactured from 316l stainless steel with electropolish finish. Includes polypropylene cap with 1/4" white compression fitting and white silicone gasket. .040 inch hole for aeration of fermentation cultures where shear force is less critical. MADE IN THE USA. Specifications:316 L Stainless SteelRa 32 Finish or Better
Catalog No. 50-194-4804 Supplier Bellco Glass, Inc. Supplier No. 196452150
Sparging accessory for vertical sidearm spinner flasks. Manufactured from 316l stainless steel with electropolish finish. Includes polypropylene cap with 1/4" white compression fitting and white silicone gasket. .040 inch hole for aeration of fermentation cultures where shear force is less critical. MADE IN THE USA. Specifications:316 L Stainless SteelRa 32 Finish or Better