Learn More
Bellco Glass, Inc. L-Bubbler Tube,15L With (10) .040 Holes"
SDP
Catalog No. 501944774
Encompass_Preferred
Manufactured from 316l stainless steel. Supplied with cap and compression fitting. MADE IN THE USA Ra 32 finish or better Dimensions: 18 1/2" x 4 3/4" x.250" tube with hose barb end .040 holes Includes: polypropylene cap and compression fitting
Catalog No. 50-194-4774 Supplier Bellco Glass, Inc. Supplier No. 196415145
Manufactured from 316l stainless steel. Supplied with cap and compression fitting. MADE IN THE USA Ra 32 finish or better Dimensions: 18 1/2" x 4 3/4" x.250" tube with hose barb end .040 holes Includes: polypropylene cap and compression fitting