Learn More
Bellco Glass, Inc. L-Bubbler Tube W/Frit, 15L
SDP
Catalog No. 501944772
Encompass_Preferred
Manufactured from 316l electropolished 1/4" stainless steel with hose barb. Ideal use for sparging of vertical sidearm spinner flasks. Includes polypropylene cap with 1/4" compression fitting and white silicone gasket. 10 micron porosity frit for cell cultures where shear is critical. Includes pp cap with 1/4" compression fitting and white silicone gasket. MADE IN THE USA. DIMENSIONS: 20 X 5 1/8"
Catalog No. 50-194-4772 Supplier Bellco Glass, Inc. Supplier No. 196415045
Manufactured from 316l electropolished 1/4" stainless steel with hose barb. Ideal use for sparging of vertical sidearm spinner flasks. Includes polypropylene cap with 1/4" compression fitting and white silicone gasket. 10 micron porosity frit for cell cultures where shear is critical. Includes pp cap with 1/4" compression fitting and white silicone gasket. MADE IN THE USA. DIMENSIONS: 20 X 5 1/8"