Learn More
Bellco Glass, Inc. L-Bubbler Tube w/frit,8L
SDP
Catalog No. 501944871
Encompass_Preferred
Sparging accessory for vertical sidearm spinner flasks316L electropolished SST, 1/4" OD; 10micron porosity frit.040" holes for aeration of fermentation cultures where shear force is less critical. MADE IN THE USA. Includes PP cap with 1/4" white compression fitting and white silicone gasketDIMENSIONS: 18 1/2 X 4 4/2"
Catalog No. 50-194-4871 Supplier Bellco Glass, Inc. Supplier No. 196480045
Sparging accessory for vertical sidearm spinner flasks316L electropolished SST, 1/4" OD; 10micron porosity frit.040" holes for aeration of fermentation cultures where shear force is less critical. MADE IN THE USA. Includes PP cap with 1/4" white compression fitting and white silicone gasketDIMENSIONS: 18 1/2 X 4 4/2"