Learn More
Bellco Glass, Inc. L-Bubbler Tube w/holes,8L (10) .040" holes
SDP
Catalog No. 501944872
Encompass_Preferred
L-bubbler tube with drilled holes for 8l ohd vessel. MADE IN THE USA. (10) .040" holes for spargingSupplied with cap & comp. FittingDimensions: 18 1/2" x 4 3/4" x .250" od tube with hose barb end316l SST material
Catalog No. 50-194-4872 Supplier Bellco Glass, Inc. Supplier No. 196481045
L-bubbler tube with drilled holes for 8l ohd vessel. MADE IN THE USA. (10) .040" holes for spargingSupplied with cap & comp. FittingDimensions: 18 1/2" x 4 3/4" x .250" od tube with hose barb end316l SST material