Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Buthionine-(S,R)-sulfoximine | 83730-53-4 | MFCD00067000 | 50mg
SDP

Catalog No. 501872147
Encompass_Preferred

L-Buthionine-(S,R)-sulfoximine | Purity: >98.0% | MW: 222.31 | 83730-53-4 | MFCD00067000 | 50mg

Catalog No. 50-187-2147 Supplier Medchemexpress LLC Supplier No. HY106376A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Buthionine-(S,R)-sulfoximine | Purity: >98.0% | MW: 222.31 | 83730-53-4 | MFCD00067000 | 50mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.