Learn More
Medchemexpress LLC L-buthionine-(S,R)-sulfoximine | 83730-53-4 | MFCD00067000 | 99.4% | 222.31 g/mol | C8H18N2O3S | 100 MG
SDP
Catalog No. 5000243256
Encompass_Preferred
L-buthionine-(S,R)-sulfoximine (L-BSO) is a small-molecule inhibitor of glutathione biosynthesis used in biochemical and cell-based research to deplete intracellular glutathione and probe oxidative stress, redox signaling, and drug-resistance mechanisms.
  • Inhibits glutathione synthesis to induce oxidative stress in cells.
  • Useful for studying redox signaling, drug resistance, and apoptosis.
  • Supplied as a high-purity solid suitable for biochemical assays.
  • Available in multiple small-pack quantities for research use.
  • Compatible with common solvents for in vitro and cell culture applications.
Catalog No. 50-002-43256 Supplier Medchemexpress LLC Supplier No. HY106376A100MG
L-buthionine-(S,R)-sulfoximine (L-BSO) is a small-molecule inhibitor of glutathione biosynthesis used in biochemical and cell-based research to deplete intracellular glutathione and probe oxidative stress, redox signaling, and drug-resistance mechanisms.
  • Inhibits glutathione synthesis to induce oxidative stress in cells.
  • Useful for studying redox signaling, drug resistance, and apoptosis.
  • Supplied as a high-purity solid suitable for biochemical assays.
  • Available in multiple small-pack quantities for research use.
  • Compatible with common solvents for in vitro and cell culture applications.