Learn More
Medchemexpress LLC L-buthionine-(S,R)-sulfoximine hydrochloride | 00-00-0 | MFCD00067000 | 258.77 | C8H19ClN2O3S | 50 MG
SDP
Catalog No. 5000242172
Encompass_Preferred
L-Buthionine-(S,R)-sulfoximine hydrochloride is a cell-permeable, potent, and irreversible inhibitor of γ-glutamylcysteine synthetase used in research to deplete intracellular glutathione and investigate oxidative stress and ferroptosis pathways.
  • Potent, irreversible inhibitor of γ-glutamylcysteine synthetase.
  • Cell-permeable and fast acting.
  • Depletes intracellular glutathione for studies of oxidative stress and ferroptosis.
  • High aqueous solubility: H2O 70 mg/mL (ultrasonic may be required).
  • Recommended storage: sealed, away from moisture; in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-42172 Supplier Medchemexpress LLC Supplier No. HY106376C50MG
L-Buthionine-(S,R)-sulfoximine hydrochloride is a cell-permeable, potent, and irreversible inhibitor of γ-glutamylcysteine synthetase used in research to deplete intracellular glutathione and investigate oxidative stress and ferroptosis pathways.
  • Potent, irreversible inhibitor of γ-glutamylcysteine synthetase.
  • Cell-permeable and fast acting.
  • Depletes intracellular glutathione for studies of oxidative stress and ferroptosis.
  • High aqueous solubility: H2O 70 mg/mL (ultrasonic may be required).
  • Recommended storage: sealed, away from moisture; in solvent -80°C (6 months) or -20°C (1 month).