Learn More
AdipoGen L-(-)-Camphoric acid (1 g)
SDP
Supplier:  AdipoGen AGCN20456G001
Encompass_Preferred
Chemical. CAS: 560-09-8. Formula: C10H16O4. MW: 200.2. Semi-synthetic. Chirality inducing agent. Useful tool as resolving agent to separate the isomers and purify racemic mixtures (e.g. DL-lysine) by fractionally crystallizing the product to obtain the pure isomer (e.g. D-lysine). Can also be used as building block.
Catalog No. 50-205-3624