Learn More
Medchemexpress LLC L-Canavanine sulfate | 2219-31-0 | 99.4% | 10 MM 1 ML
SDP
Catalog No. 5000413278
Encompass_Preferred
L-Canavanine sulfate is a selective inhibitor of inducible NO synthase, exhibiting low cytotoxicity in normal cells in vitro while potentiating the effects of certain chemotherapy drugs in cancer cells. In vivo, it can help restore blood pressure in endotoxic shock models and has shown antitumor activity.
  • Selective inhibitor of inducible NO synthase.
  • Low cytotoxicity in normal cells in vitro.
  • Potentiates the cytotoxicity of certain chemotherapy drugs in cancer cells.
  • Helps restore blood pressure in endotoxic shock.
  • Exhibits antitumor activity.
Catalog No. 50-004-13278 Supplier Medchemexpress LLC Supplier No. HYB1581A10MM/1ML
L-Canavanine sulfate is a selective inhibitor of inducible NO synthase, exhibiting low cytotoxicity in normal cells in vitro while potentiating the effects of certain chemotherapy drugs in cancer cells. In vivo, it can help restore blood pressure in endotoxic shock models and has shown antitumor activity.
  • Selective inhibitor of inducible NO synthase.
  • Low cytotoxicity in normal cells in vitro.
  • Potentiates the cytotoxicity of certain chemotherapy drugs in cancer cells.
  • Helps restore blood pressure in endotoxic shock.
  • Exhibits antitumor activity.