Learn More
Cayman Chemical L-Carnitine-d3 (chloride), Purity: ≥99% deuterated forms (d1-d3), CAS: 350818-62-1, 25 mg
SDP
Catalog No. 502935139
Encompass_Preferred
An internal standard for the quantification of L-carnitine by GC- or LC-MS
Catalog No. 50-293-5139 Supplier Cayman Chemical Supplier No. 2656525 MG
An internal standard for the quantification of L-carnitine by GC- or LC-MS