Learn More
Cayman Chemical L-Carnitine-d3 (chloride), Purity: ≥99% deuterated forms (d1-d3), CAS: 350818-62-1, 5 mg
SDP
Catalog No. 502933847
Encompass_Preferred
An internal standard for the quantification of L-carnitine by GC- or LC-MS
Catalog No. 50-293-3847 Supplier Cayman Chemical Supplier No. 265655 MG
An internal standard for the quantification of L-carnitine by GC- or LC-MS