Learn More
Medchemexpress LLC L-Carnitine | 541-15-1 | >99.96 % | 10 G
SDP
Catalog No. 5000407149
Encompass_Preferred
L-Carnitine ((R)-Carnitine), a highly polar, small zwitterion, is an essential co-factor for the mitochondrial β-oxidation pathway. It functions to transport long chain fatty acyl-CoAs into the mitochondria for degradation by β-oxidation. L-Carnitine is also an antioxidant and can ameliorate metabolic imbalances in many inborn errors of metabolism.
  • Highly polar, small zwitterion
  • Essential co-factor for mitochondrial β-oxidation pathway
  • Transports long chain fatty acyl-CoAs into mitochondria
  • Antioxidant properties
  • Ameliorates metabolic imbalances in inborn errors of metabolism
Catalog No. 50-004-07149 Supplier Medchemexpress LLC Supplier No. HYB039910G
L-Carnitine ((R)-Carnitine), a highly polar, small zwitterion, is an essential co-factor for the mitochondrial β-oxidation pathway. It functions to transport long chain fatty acyl-CoAs into the mitochondria for degradation by β-oxidation. L-Carnitine is also an antioxidant and can ameliorate metabolic imbalances in many inborn errors of metabolism.
  • Highly polar, small zwitterion
  • Essential co-factor for mitochondrial β-oxidation pathway
  • Transports long chain fatty acyl-CoAs into mitochondria
  • Antioxidant properties
  • Ameliorates metabolic imbalances in inborn errors of metabolism