Learn More
Medchemexpress LLC L-Carnitine | 541-15-1 | MFCD00038747 | 100.0% | 50 G
SDP
Catalog No. 5000406558
Encompass_Preferred
L-Carnitine, also known as (R)-Carnitine or Levocarnitine, is a highly polar, small zwitterion that serves as an essential co-factor for the mitochondrial β-oxidation pathway. Its primary function involves transporting long-chain fatty acyl-CoAs into the mitochondria for degradation through β-oxidation. L-Carnitine also acts as an antioxidant and has demonstrated the ability to ameliorate metabolic imbalances in various inborn errors of metabolism. This product is intended for research use only.
  • Essential co-factor for mitochondrial β-oxidation
  • Transports long-chain fatty acyl-CoAs into mitochondria
  • Acts as an antioxidant
  • Ameliorates metabolic imbalances
  • For research use only
Catalog No. 50-004-06558 Supplier Medchemexpress LLC Supplier No. HYB039950G
L-Carnitine, also known as (R)-Carnitine or Levocarnitine, is a highly polar, small zwitterion that serves as an essential co-factor for the mitochondrial β-oxidation pathway. Its primary function involves transporting long-chain fatty acyl-CoAs into the mitochondria for degradation through β-oxidation. L-Carnitine also acts as an antioxidant and has demonstrated the ability to ameliorate metabolic imbalances in various inborn errors of metabolism. This product is intended for research use only.
  • Essential co-factor for mitochondrial β-oxidation
  • Transports long-chain fatty acyl-CoAs into mitochondria
  • Acts as an antioxidant
  • Ameliorates metabolic imbalances
  • For research use only