Learn More
Medchemexpress LLC HY-B0399 5g , L-Carnitine CAS:541-15-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB03995G
Encompass_Preferred
HY-B0399 5g L-Carnitine CAS: 541-15-1 L-Carnitine ((R)-Carnitine), a highly polar, small zwitterion, is an essential co-factor for the mitochondrial β-oxidation pathway. L-Carnitine functions to transport long chain fatty acyl-CoAs into the mitochondria for degradation by β-oxidation. L-Carnitine is an antioxidant. L-Carnitine hydrochloride can ameliorate metabolic imbalances in many inborn errors of metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0292