Learn More
Medchemexpress LLC L-Carnitine-d9 | 126827-79-0 | ≥99.0% | 1 MG
SDP
Catalog No. 5000407151
Encompass_Preferred
L-Carnitine-d9 is a deuterium-labeled L-Carnitine, an endogenous molecule involved in fatty acid metabolism. Biosynthesized in the human body using L-lysine and L-methionine, it transports long-chain fatty acyl-CoAs into mitochondria for degradation through β-oxidation. It can also improve metabolic imbalances in various inborn errors of metabolism.
  • Deuterium-labeled L-Carnitine for tracing during drug development.
  • Participates in fatty acid metabolism.
  • Transports long-chain fatty acyl-CoAs into mitochondria.
  • May improve metabolic imbalances in inborn errors of metabolism.
Catalog No. 50-004-07151 Supplier Medchemexpress LLC Supplier No. HYB0399S1MG
L-Carnitine-d9 is a deuterium-labeled L-Carnitine, an endogenous molecule involved in fatty acid metabolism. Biosynthesized in the human body using L-lysine and L-methionine, it transports long-chain fatty acyl-CoAs into mitochondria for degradation through β-oxidation. It can also improve metabolic imbalances in various inborn errors of metabolism.
  • Deuterium-labeled L-Carnitine for tracing during drug development.
  • Participates in fatty acid metabolism.
  • Transports long-chain fatty acyl-CoAs into mitochondria.
  • May improve metabolic imbalances in inborn errors of metabolism.