Learn More
Apexbio Technology LLC L-CARNITINE-D9-1MG
SDP
Catalog No. 5000570837
Encompass_Preferred
L-Carnitine-d9 (CAS No 126827-79-0) is a deuterated form of L-carnitine commonly used in biomedical research to study metabolic processes Originating as an amino acid derivative it plays a key role in the mitochondrial oxidation of long-chain fatty acids by facilitating their transport into the mitochondria This compound targets pathways critical to energy production often investigated in vitro within the nanomolar to low micromolar range Typical applications include studies in cell models and animal studies to explore metabolic disorders and enhance drug screening methodologies Researchers utilize L-Carnitine-d9 to understand better the intricate dynamics of fatty acid metabolism employing experimental concentrations that are tailored to the specific requirements of each study
Catalog No. 50-005-70837 Supplier Apexbio Technology LLC Supplier No. SL10111
L-Carnitine-d9 (CAS No 126827-79-0) is a deuterated form of L-carnitine commonly used in biomedical research to study metabolic processes Originating as an amino acid derivative it plays a key role in the mitochondrial oxidation of long-chain fatty acids by facilitating their transport into the mitochondria This compound targets pathways critical to energy production often investigated in vitro within the nanomolar to low micromolar range Typical applications include studies in cell models and animal studies to explore metabolic disorders and enhance drug screening methodologies Researchers utilize L-Carnitine-d9 to understand better the intricate dynamics of fatty acid metabolism employing experimental concentrations that are tailored to the specific requirements of each study