Learn More
Medchemexpress LLC L-Carnitine-d9 chloride | 2687961-04-0 | 99.0% | 5 MG
SDP
Catalog No. 5000411032
Encompass_Preferred
L-Carnitine-d9 chloride is a deuterium-labeled L-Carnitine chloride. This highly polar, small zwitterion serves as an essential co-factor for the mitochondrial β-oxidation pathway, facilitating the transport of long-chain fatty acyl-CoAs into mitochondria for degradation. It also functions as an antioxidant and can alleviate metabolic imbalances in various inborn errors of metabolism.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration offers potential to influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11032 Supplier Medchemexpress LLC Supplier No. HYB2246S5MG
L-Carnitine-d9 chloride is a deuterium-labeled L-Carnitine chloride. This highly polar, small zwitterion serves as an essential co-factor for the mitochondrial β-oxidation pathway, facilitating the transport of long-chain fatty acyl-CoAs into mitochondria for degradation. It also functions as an antioxidant and can alleviate metabolic imbalances in various inborn errors of metabolism.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration offers potential to influence pharmacokinetic and metabolic profiles of drugs.