Learn More
Selleck Chemical LLC L-Carnitine hydrochloride
Supplier: Selleck Chemical LLC S592225MG
L-Carnitine ((R)-Carnitine Levocarnitine) hydrochloride improves oxygen saturation and inhibits leukotriene synthesis L-Carnitine hydrochloride functions to transport long chain fatty acyl-CoAs into the mitochondria for degradation by -oxidation