Learn More
Selleck Chemical LLC L-Carnitine hydrochloride
SDP
Catalog No. 5000003938
Encompass_Preferred
L-Carnitine ((R)-Carnitine Levocarnitine) hydrochloride improves oxygen saturation and inhibits leukotriene synthesis L-Carnitine hydrochloride functions to transport long chain fatty acyl-CoAs into the mitochondria for degradation by -oxidation
Catalog No. 50-000-03938 Supplier Selleck Chemical LLC Supplier No. S59221G
L-Carnitine ((R)-Carnitine Levocarnitine) hydrochloride improves oxygen saturation and inhibits leukotriene synthesis L-Carnitine hydrochloride functions to transport long chain fatty acyl-CoAs into the mitochondria for degradation by -oxidation