Learn More
Medchemexpress LLC L-Carnitine hydrochloride | 6645-46-1 | 99.88% | 50 MG
SDP
Catalog No. 5000410513
Encompass_Preferred
L-Carnitine hydrochloride is a highly polar, small zwitterion that acts as an essential co-factor for the mitochondrial β-oxidation pathway. It transports long-chain fatty acyl-CoAs into the mitochondria for degradation by β-oxidation. It also functions as an antioxidant and can improve metabolic imbalances in various inborn errors of metabolism.
  • Transports long-chain fatty acyl-CoAs into mitochondria
  • Degrades fatty acids by β-oxidation
  • Acts as an antioxidant
  • Ameliorates metabolic imbalances in many inborn errors of metabolism
Catalog No. 50-004-10513 Supplier Medchemexpress LLC Supplier No. HYB224650MG
L-Carnitine hydrochloride is a highly polar, small zwitterion that acts as an essential co-factor for the mitochondrial β-oxidation pathway. It transports long-chain fatty acyl-CoAs into the mitochondria for degradation by β-oxidation. It also functions as an antioxidant and can improve metabolic imbalances in various inborn errors of metabolism.
  • Transports long-chain fatty acyl-CoAs into mitochondria
  • Degrades fatty acids by β-oxidation
  • Acts as an antioxidant
  • Ameliorates metabolic imbalances in many inborn errors of metabolism