Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Carnitine hydrochloride, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015116 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5116 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5116 Supplier MilliporeSigma Supelco Supplier No. 9495450MG
Add to Cart
Add to Cart

Specifications

CAS 6645-46-1
Linear Formula (CH3)3 N+CH2 CH(OH)CH2 CO2 H · Cl-
MDL Number MFCD00066100
Synonym (-)-beta-Hydroxy-gamma-(trimethylammonio)butyrate; Vitamin BT
Type Analytical Standard
Quantity 50 mg
Formula Weight 197.66
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.