Learn More
Medchemexpress LLC L-Carnitine tartrat 25g
SDP
Catalog No. 5000206838
Encompass_Preferred
L-Carnitine tartrate is a highly polar small zwitterion L-Carnitine tartrate is an essential co-factor for the mitochondrial -oxidation pathway L-Carnitine tartrate functions to transport long chain fatty acyl-CoAs into the mitochondria for degradation by -oxidation L-Carnitine tartrate is an antioxidant L-Carnitine tartrate can ameliorate metabolic imbalances in many inborn errors of metabolism[1][2][3]
Catalog No. 50-002-06838 Supplier Medchemexpress LLC Supplier No. 25GHYW105518
L-Carnitine tartrate is a highly polar small zwitterion L-Carnitine tartrate is an essential co-factor for the mitochondrial -oxidation pathway L-Carnitine tartrate functions to transport long chain fatty acyl-CoAs into the mitochondria for degradation by -oxidation L-Carnitine tartrate is an antioxidant L-Carnitine tartrate can ameliorate metabolic imbalances in many inborn errors of metabolism[1][2][3]