Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Carnitine-(trimethyl-d9) inner salt, Analytical Standard, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012874 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2874 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2874 Supplier MilliporeSigma Supelco Supplier No. 9368910MG
Add to Cart
Add to Cart

Specifications

CAS 126827-79-0
Linear Formula C7D9H6NO3
Synonym (R)-3-Carboxy-2-hydroxy-N,N,N-tri(methyl-d3)-1-propanaminium inner salt
Quantity 10 mg
Formula Weight 170.25
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.