Learn More
Cayman Chemical L-Carnosine-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2714339-90-7, 1 mg
SDP
Catalog No. 502936730
Encompass_Preferred
An internal standard for the quantification of L-carnosine by GC- or LC-MS
Catalog No. 50-293-6730 Supplier Cayman Chemical Supplier No. 267811 MG
An internal standard for the quantification of L-carnosine by GC- or LC-MS