Learn More
Cayman Chemical L-CarnosIn-d4 500ug
SDP
Catalog No. 502932939
Encompass_Preferred
An internal standard for the quantification of L-carnosine by GC- or LC-MS
Catalog No. 50-293-2939 Supplier Cayman Chemical Supplier No. 26781500 UG
An internal standard for the quantification of L-carnosine by GC- or LC-MS