Learn More
Medchemexpress LLC L-Carnosine | 305-84-0 | 50 MG
SDP
Catalog No. 5000454232
Encompass_Preferred
L-Carnosine is an analytical standard of L-Carnosine, intended for research and analytical applications. It is a dipeptide composed of beta-alanine and histidine, found in human brain, muscle, and gastrointestinal tissues. It acts as an endogenous metabolite and possesses anti-inflammatory and neuroprotective properties.
  • Functions as a non-enzymatic free radical scavenger
  • Acts as a natural antioxidant
  • Inhibits biochemical changes associated with aging
  • Exhibits antioxidant effects through metal ion chelation
  • Demonstrates superoxide dismutase (SOD)-like activity
  • Scavenges reactive oxygen species (ROS) and free radicals
Catalog No. 50-004-54232 Supplier Medchemexpress LLC Supplier No. HYW013494R50MG
L-Carnosine is an analytical standard of L-Carnosine, intended for research and analytical applications. It is a dipeptide composed of beta-alanine and histidine, found in human brain, muscle, and gastrointestinal tissues. It acts as an endogenous metabolite and possesses anti-inflammatory and neuroprotective properties.
  • Functions as a non-enzymatic free radical scavenger
  • Acts as a natural antioxidant
  • Inhibits biochemical changes associated with aging
  • Exhibits antioxidant effects through metal ion chelation
  • Demonstrates superoxide dismutase (SOD)-like activity
  • Scavenges reactive oxygen species (ROS) and free radicals