Learn More
Medchemexpress LLC L-Carnosine | 305-84-0 | 99.94% | 226.24 | 10 MG
SDP
Catalog No. 5000454564
Encompass_Preferred
L-Carnosine is a dipeptide composed of beta-alanine and histidine, naturally occurring in human brain, muscle, and gastrointestinal tissues. This endogenous metabolite acts as a non-enzymatic free radical scavenger and a natural antioxidant with anti-inflammatory and neuroprotective properties. It can inhibit biochemical changes associated with aging, attributed to its chelating effect against metal ions, superoxide dismutase (SOD)-like activity, and its ability to scavenge reactive oxygen species (ROS) and free radicals.
  • Acts as a non-enzymatic free radical scavenger
  • Functions as a natural antioxidant
  • Exhibits anti-inflammatory properties
  • Provides neuroprotective benefits
  • Inhibits biochemical changes associated with aging
  • Chelates metal ions
  • Scavenges reactive oxygen species and free radicals
Catalog No. 50-004-54564 Supplier Medchemexpress LLC Supplier No. HYW013494R10MG
L-Carnosine is a dipeptide composed of beta-alanine and histidine, naturally occurring in human brain, muscle, and gastrointestinal tissues. This endogenous metabolite acts as a non-enzymatic free radical scavenger and a natural antioxidant with anti-inflammatory and neuroprotective properties. It can inhibit biochemical changes associated with aging, attributed to its chelating effect against metal ions, superoxide dismutase (SOD)-like activity, and its ability to scavenge reactive oxygen species (ROS) and free radicals.
  • Acts as a non-enzymatic free radical scavenger
  • Functions as a natural antioxidant
  • Exhibits anti-inflammatory properties
  • Provides neuroprotective benefits
  • Inhibits biochemical changes associated with aging
  • Chelates metal ions
  • Scavenges reactive oxygen species and free radicals